5-Chloro-2-methoxybenzaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H31860-06
H31860-14
AAH31860-06EA
86.9
USD
AAH31860-06
AAH31860-14
5-Chloro-2-methoxybenzaldehyde 97%
5-Chloro-2-methoxybenzaldehyde
Purity: 97%
CAS No.: 7035-09-8
Molecular Formula: C8H7ClO2
Formula Weight: 170.60
Odor: Characteristic
Melting Point: 75-79°
MDL No.: MFCD00033854
Formula:
C₈H₇ClO₂ MW: 170.60 g/mol Melting Pt: 75…79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00033854 CAS Number: 7035-09-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...