5-(Trifluoromethyl)pyridine-2-carboxaldehyde 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
5-(Trifluoromethyl)picolinaldehyde
H31858-03
H31858-MD
AAH31858-03EA
456
USD
AAH31858-03
AAH31858-MD
5-(Trifluoromethyl)pyridine-2-carboxaldehyde 95%
5-(Trifluoromethyl)pyridine-2-carboxaldehyde
Purity: 95%
CAS No.: 31224-82-5
Molecular Formula: C7H4F3NO
Formula Weight: 175.11
Storage Temperature: 2°C to 8°C
Melting Point: 31-33°
MDL No.: MFCD07375388
Formula:
C₇H₄F₃NO Melting Pt: 31…33 °C Storage Temperature: Ambient |
MDL Number:
MFCD07375388 CAS Number: 31224-82-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...