Thionyl chloride 1 M in dichloromethane
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Thionyl dichloride
H31857-AE
H31857-AP
AAH31857-AEEA
103.86
USD
AAH31857-AE
AAH31857-AP
Thionyl chloride 1 M in dichloromethane
Thionyl chloride
CAS No.: 7719-09-7
Molecular Formula: Cl2OS
Formula Weight: 118.97
UN No.: 2922
MDL No.: MFCD00011449
Formula:
Cl₂OS Density: 1.35 Storage Temperature: Ambient |
MDL Number:
MFCD00011449 CAS Number: 7719-09-7 EINECS: 231-748-8 UN: 2922 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...