2-Iodo-1-(2-tetrahydropyranyl)-1H-imidazole 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H31846-03
H31846-MD
AAH31846-03EA
201
USD
AAH31846-03
AAH31846-MD
2-Iodo-1-(2-tetrahydropyranyl)-1H-imidazole 95%
2-Iodo-1-(2-tetrahydropyranyl)-1H-imidazole
Purity: 95%
Molecular Formula: C8H11IN2O
Formula Weight: 278.09
MDL No.: MFCD11100955
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...