4-Bromo-2-methylbenzenesulfonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H31835-03
H31835-09
AAH31835-03EA
20.6
USD
AAH31835-03
AAH31835-09
4-Bromo-2-methylbenzenesulfonyl chloride 97%
4-Bromo-2-methylbenzenesulphonyl chloride
Purity: 97%
CAS No.: 139937-37-4
Molecular Formula: C7H6BrClO2S
Formula Weight: 269.55
Melting Point: 63-65°
Boiling Point: 164°/15mm
UN No.: 3265
MDL No.: MFCD01365820
Formula:
C₇H₆BrClO₂S Boiling Pt: 164 °C/15 mm Melting Pt: 63…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00219927 CAS Number: 139937-37-4 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...