Titanium(IV) chloride 1 M in dichloromethane
Supplier: Thermo Scientific Chemicals
|
Danger
|
H31830-AE
H31830-AP
AAH31830-AEEA
63.51
USD
AAH31830-AE
AAH31830-AP
Titanium(IV) chloride 1 M in dichloromethane
Titanium(IV) chloride
CAS No.: 7550-45-0
Molecular Formula: Cl4Ti
Formula Weight: 189.69
UN No.: 2922
MDL No.: MFCD00011267
Formula:
Cl₄Ti Density: 1.365 Storage Temperature: Ambient |
MDL Number:
MFCD00011267 CAS Number: 7550-45-0 EINECS: 231-441-9 UN: 2922 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...