4-(2-Thiazolyl)phenol 97%

Supplier: Thermo Scientific Chemicals
Warning

H31827-03 H31827-09
AAH31827-03EA 27.4 USD
AAH31827-03 AAH31827-09
4-(2-Thiazolyl)phenol 97%
4-(2-Thiazolyl)phenol

Purity: 97% CAS No.: 81015-49-8 Molecular Formula: C9H7NOS Formula Weight: 177.28 Melting Point: 162-165° MDL No.: MFCD07368615

Formula: C₉H₇NOS
MW: 177.28 g/mol
Melting Pt: 162…165 °C
Storage Temperature: Ambient
MDL Number: MFCD07368615
CAS Number: 81015-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR