3-Ethoxy-2,4-difluorobenzoic acid 97%, JRD

Supplier: Thermo Scientific Chemicals
Warning

H31812-06 H31812-03
AAH31812-06EA 349 USD
AAH31812-06 AAH31812-03
3-Ethoxy-2,4-difluorobenzoic acid 97%, JRD
3-Ethoxy-2,4-difluorobenzoic acid

Purity: 97% Molecular Formula: C9H8F2O3 Formula Weight: 202.15 Melting Point: 134-136° MDL No.: MFCD09258723

Formula: C₉H₈F₂O₃
MW: 202.15 g/mol
Melting Pt: 134…136 °C
Storage Temperature: Ambient
MDL Number: MFCD09258723
CAS Number: 1017779-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR