6-Bromopicolinamide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Bromopyridine-2-carboxamide

H31811-03 H31811-06
AAH31811-03EA 57.8 USD
AAH31811-03 AAH31811-06
6-Bromopicolinamide 97%
6-Bromopicolinamide

Purity: 97% CAS No.: 25194-52-9 Molecular Formula: C6H5BrN2O Formula Weight: 201.02 MDL No.: MFCD00234154

Formula: C₆H₅BrN₂O
MW: 201.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234154
CAS Number: 25194-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR