Ethyl-2-aminothiazole-5-carboxylate 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Ethyl 2-aminothiazole-5-carboxylate
H31769-03
H31769-06
AAH31769-03EA
40.2
USD
AAH31769-03
AAH31769-06
Ethyl-2-aminothiazole-5-carboxylate 97%
Ethyl-2-aminothiazole-5-carboxylate
Purity: 97%
CAS No.: 32955-21-8
Molecular Formula: C6H8N2O2S
Formula Weight: 172.21
Storage Temperature: 2°C to 8°C
Melting Point: 158-160°
MDL No.: MFCD00602139
Formula:
C₆H₈N₂O₂S Melting Pt: 158…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00602139 CAS Number: 32955-21-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...