3-Fluoro-4-methoxybenzyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Fluoro-4-methoxybenzylchloride
H31756-03
H31756-06
AAH31756-03EA
71.1
USD
AAH31756-03
AAH31756-06
3-Fluoro-4-methoxybenzyl chloride 98%
3-Fluoro-4-methoxybenzyl chloride
Purity: 98%
CAS No.: 351-52-0
Molecular Formula: C8H8ClFO
Formula Weight: 174.60
Melting Point: 35-39°
UN No.: 3261
MDL No.: MFCD04973773
Formula:
C₈H₈ClFO Melting Pt: 35…39 °C Storage Temperature: Ambient |
MDL Number:
MFCD04973773 CAS Number: 351-52-0 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...