Pyrrolidin-2-ylacetic acid hydrochloride 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Pyrrolidin-2-yl-acetic acid hydrochloride

H31749-03 H31749-MD
AAH31749-03EA 245 USD
AAH31749-03 AAH31749-MD
Pyrrolidin-2-ylacetic acid hydrochloride 97%
Pyrrolidin-2-ylacetic acid hydrochloride

Purity: 97% CAS No.: 71985-79-0 Molecular Formula: C6H11NO2.HCl Formula Weight: 165.62 Melting Point: 171-174° MDL No.: MFCD11506301

Formula: C6H12ClNO2
MW: 165.62 g/mol
Melting Pt: 171…174 °C
Storage Temperature: Ambient
MDL Number: MFCD11506301
CAS Number: 71985-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR