4-Isopropenyltoluene stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4,alpha-Dimethylstyrene, 1-Isopropenyl-4-methylbenzene

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

H31739-06 H31739-14 H31739-22
AAH31739-06EA 65.6 USD
AAH31739-06 AAH31739-14 AAH31739-22
4-Isopropenyltoluene stabilized
4-Isopropenyltoluene

Stab. with 4-tert-butylcatechol


CAS No.: 1195-32-0 Molecular Formula: C10H12 Formula Weight: 132.21 Flash Point: 63°(145°F) Refractive Index: 1.5090 Boiling Point: 186-189° MDL No.: MFCD00036510

Formula: C₁₀H₁₂
MW: 132.21 g/mol
Boiling Pt: 186…189 °C
Density: 0.872
Flash Pt: 63 °C(145 °F)
Storage Temperature: Ambient
MDL Number: MFCD00036510
CAS Number: 1195-32-0
EINECS: 214-795-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR