Celatom® FW-14

Supplier: Thermo Scientific Chemicals

H31733-0B H31733-30
AAH31733-0BEA 127.72 USD
AAH31733-0B AAH31733-30
Celatom® FW-14
Celatom® FW-14

CAS No.: 68855-54-9 MDL No.: MFCD00132803

Storage Temperature: Ambient
MDL Number: MFCD00132803
CAS Number: 68855-54-9
EINECS: 272-489-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR