4-Bromo-2-fluoro-5-(trifluoromethyl)aniline 97%

Supplier: Thermo Scientific Chemicals
Warning

H31727-03 H31727-09
AAH31727-03EA 21.1 USD
AAH31727-03 AAH31727-09
4-Bromo-2-fluoro-5-(trifluoromethyl)aniline 97%
4-Bromo-2-fluoro-5-(trifluoromethyl)aniline

Purity: 97% CAS No.: 104460-70-0 Molecular Formula: C7H4BrF4N Formula Weight: 258.01 Melting Point: 30-41° UN No.: 2811 MDL No.: MFCD04973757

Formula: C₇H₄BrF₄N
MW: 258.01 g/mol
Melting Pt: 30…41 °C
Storage Temperature: Ambient
MDL Number: MFCD04973757
CAS Number: 104460-70-0
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR