2,4-Dimethoxybenzenesulfonamide 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H31707-03
H31707-06
AAH31707-03EA
50.9
USD
AAH31707-03
AAH31707-06
2,4-Dimethoxybenzenesulfonamide 96%
2,4-Dimethoxybenzenesulphonamide
Purity: 96%
CAS No.: 51770-71-9
Molecular Formula: C8H11NO4S
Formula Weight: 217.25
Melting Point: 166-170°
MDL No.: MFCD06147001
Formula:
C₈H₁₁NO₄S Melting Pt: 166…170 °C Storage Temperature: Ambient |
MDL Number:
MFCD06147001 CAS Number: 51770-71-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...