4-(4-Bromophenoxy)benzaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-(4-Bromo-phenoxy)benzaldehyde
H31702-03
H31702-06
AAH31702-03EA
71.8
USD
AAH31702-03
AAH31702-06
4-(4-Bromophenoxy)benzaldehyde 97%
4-(4-Bromophenoxy)benzaldehyde
Purity: 97%
CAS No.: 69240-56-8
Molecular Formula: C13H9BrO2
Formula Weight: 277.12
Melting Point: 69-73°
UN No.: 3077
MDL No.: MFCD08276811
Formula:
C₁₃H₉BrO₂ Melting Pt: 69…73 °C Storage Temperature: Ambient |
MDL Number:
MFCD08276811 CAS Number: 69240-56-8 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...