2-Fluoro-4-pyridone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Fluoropyridin-4-ol, 2-Fluoro-4-hydroxypyridine
H31697-03
H31697-MD
AAH31697-03EA
253
USD
AAH31697-03
AAH31697-MD
2-Fluoro-4-pyridone 97%
2-Fluoro-4-pyridone
Purity: 97%
CAS No.: 22282-69-5
Molecular Formula: C5H4FNO
Formula Weight: 113.09
Melting Point: 146-149°
MDL No.: MFCD04114159
Formula:
C₅H₄FNO MW: 113.09 g/mol Melting Pt: 146…149 °C Storage Temperature: Ambient |
MDL Number:
MFCD04114159 CAS Number: 22282-69-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...