4-Mercapto-4-methyl-2-pentanone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Mercapto-4-methylpentan-2-one
H31693-03
H31693-06
H31693-14
AAH31693-03EA
50.5
USD
AAH31693-03
AAH31693-06
AAH31693-14
4-Mercapto-4-methyl-2-pentanone 98%
4-Mercapto-4-methyl-2-pentanone
Purity: 98%
CAS No.: 19872-52-7
Molecular Formula: C6H12SO
Formula Weight: 132.23
Storage Temperature: -30°C to -10°C
Flash Point: 54°(129°F)
Refractive Index: 1.4620
Odor: Unpleasant
Boiling Point: 174°
UN No.: 3336
MDL No.: MFCD00085208
Formula:
C₆H₁₂OS MW: 132.23 g/mol Boiling Pt: 174 °C Density: 0.961 Flash Pt: 54 °C (129 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00085208 CAS Number: 19872-52-7 EINECS: 243-386-8 UN: 3336 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...