2,5-Dibromobenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Danger

H31689-03 H31689-06
AAH31689-03EA 56.6 USD
AAH31689-03 AAH31689-06 AAH31689-14
2,5-Dibromobenzaldehyde 97%
2,5-Dibromobenzaldehyde

Purity: 97% CAS No.: 74553-29-0 Molecular Formula: C7H4Br2O Formula Weight: 263.93 Melting Point: 88-102° MDL No.: MFCD00462864

Formula: C₇H₄Br₂O
Melting Pt: 88…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00462864
CAS Number: 74553-29-0
UN: 2923
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR