4-Hydroxy-3-(trifluoromethyl)benzonitrile 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H31639-03
H31639-06
AAH31639-03EA
137
USD
AAH31639-03
AAH31639-06
4-Hydroxy-3-(trifluoromethyl)benzonitrile 98+%
4-Hydroxy-3-(trifluoromethyl)benzonitrile
Purity: 98+%
CAS No.: 124811-71-8
Molecular Formula: C8H4F3NO
Formula Weight: 187.12
Melting Point: 171-175°
MDL No.: MFCD06797882
Formula:
C₈H₄F₃NO Melting Pt: 171…175 °C Storage Temperature: Ambient |
MDL Number:
MFCD06797882 CAS Number: 124811-71-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...