3,5-Difluoro-4-hydroxybenzonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

H31620-03 H31620-MD
AAH31620-03EA 110 USD
AAH31620-03 AAH31620-MD
3,5-Difluoro-4-hydroxybenzonitrile 97%
3,5-Difluoro-4-hydroxybenzonitrile

Purity: 97% CAS No.: 2967-54-6 Molecular Formula: C7H3F2NO Formula Weight: 155.10 Melting Point: 200-208° UN No.: 3439 MDL No.: MFCD09033181

Formula: C₇H₃F₂NO
MW: 155.1 g/mol
Melting Pt: 200…208 °C
Storage Temperature: Ambient
MDL Number: MFCD09033181
CAS Number: 2967-54-6
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR