(4-Fluorophenyl)methanesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Fluoro-α-toluenesulphonyl chloride

H31605-03 H31605-MD
AAH31605-03EA 67 USD
AAH31605-03 AAH31605-MD
(4-Fluorophenyl)methanesulfonyl chloride 97%
(4-Fluorophenyl)methanesulphonyl chloride

Purity: 97% CAS No.: 103360-04-9 Molecular Formula: C7H6ClFO2S Formula Weight: 208.64 Melting Point: 58-61° UN No.: 3261 MDL No.: MFCD01631930

Formula: C₇H₆ClFO₂S
MW: 208.64 g/mol
Melting Pt: 58…61 °C
Storage Temperature: Ambient
MDL Number: MFCD01631930
CAS Number: 103360-04-9
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR