2-Bromo-6-methoxybenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H31604-03
H31604-06
AAH31604-03EA
81.5
USD
AAH31604-03
AAH31604-06
2-Bromo-6-methoxybenzoic acid 98%
2-Bromo-6-methoxybenzoic acid
Purity: 98%
CAS No.: 31786-45-5
Molecular Formula: C8H7BrO3
Formula Weight: 231.05
MDL No.: MFCD00800732
Formula:
C₈H₇BrO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00800732 CAS Number: 31786-45-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...