3-Amino-2,6-dichloropyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

H31598-06 H31598-14
AAH31598-03EA 26.28 USD
AAH31598-03 AAH31598-06 AAH31598-14
3-Amino-2,6-dichloropyridine 97%
3-Amino-2,6-dichloropyridine

Purity: 97% CAS No.: 62476-56-6 Molecular Formula: C5H4Cl2N2 Formula Weight: 163.01 Melting Point: 120-124° MDL No.: MFCD00023417

Formula: C₅H₄Cl₂N₂
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 119 °C
Density: 1.497 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00023417
CAS Number: 62476-56-6
EINECS: 263-559-1
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR