2,3,4,6-Tetra-O-benzyl-α-D-glucopyranose 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(2S,3R,4S,5R,6R)-3,4,5-Tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-ol
H31595-03
H31595-06
AAH31595-03EA
61.6
USD
AAH31595-03
AAH31595-06
2,3,4,6-Tetra-O-benzyl-α-D-glucopyranose 98%
2,3,4,6-Tetra-O-benzyl-alpha-D-glucopyranose
Purity: 98%
CAS No.: 6564-72-3
Molecular Formula: C34H36O6
Formula Weight: 540.65
Storage Temperature: -30°C to -10°C
Melting Point: 150-151°
MDL No.: MFCD00023849
Formula:
C₃₄H₃₆O₆ MW: 540.65 g/mol Melting Pt: 150…151 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00023849 CAS Number: 6564-72-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...