4-Hydroxybenzamidine hydrochloride 95%

Supplier: Thermo Scientific Chemicals
Warning

H31566-03 H31566-09
AAH31566-03EA 28.6 USD
AAH31566-03 AAH31566-09
4-Hydroxybenzamidine hydrochloride 95%
4-Hydroxybenzamidine hydrochloride

Purity: 95% CAS No.: 38148-63-9 Molecular Formula: C7H8N2O.HCl Formula Weight: 172.61 Melting Point: 221-224° MDL No.: MFCD00136405

Formula: C₇H₉ClN₂O
Melting Pt: 221…224 °C
Storage Temperature: Ambient
MDL Number: MFCD00136405
CAS Number: 38148-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR