1-Ethoxy-1-(tributylstannyl)ethene 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tributyl(1-ethoxyvinyl)tin, Tri-n-butyl(1-ethoxyvinyl)tin, 1-Ethoxyvinyltri-n-butyltin

H31562-03 H31562-06 H31562-14
AAH31562-03EA 48.9 USD
AAH31562-03 AAH31562-06 AAH31562-14
1-Ethoxy-1-(tributylstannyl)ethene 95%
1-Ethoxy-1-(tributylstannyl)ethene

CAS No.: 97674-02-7 Molecular Formula: C16H34OSn Formula Weight: 361.14 Flash Point: 110°(230°F) Refractive Index: 1.4760 Boiling Point: 85-86°/0.07mm UN No.: 2788 MDL No.: MFCD00010240

Formula: C₁₆H₃₄OSn
MW: 361.15 g/mol
Boiling Pt: 85…86 °C/0.07 mm
Density: 1.067
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010240
CAS Number: 97674-02-7
UN: 2788
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR