2,5-Difluoro-4-hydroxybenzaldehyde 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H31528-03
H31528-06
AAH31528-03EA
127
USD
AAH31528-03
AAH31528-06
AAH31528-MD
2,5-Difluoro-4-hydroxybenzaldehyde 99%
2,5-Difluoro-4-hydroxybenzaldehyde
Purity: 99%
CAS No.: 918523-99-6
Molecular Formula: C7H4F2O2
Formula Weight: 158.11
Melting Point: 145-148°
MDL No.: MFCD04112521
Formula:
C₇H₄F₂O₂ Melting Pt: 145…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD04112521 CAS Number: 918523-99-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...