2-Bromo-3'-(trifluoromethyl)acetophenone 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H31526-03
H31526-06
H31526-09
AAH31526-03EA
109
USD
AAH31526-03
AAH31526-06
AAH31526-09
2-Bromo-3'-(trifluoromethyl)acetophenone 98%
2-Bromo-3'-(trifluoromethyl)acetophenone
Purity: 98%
CAS No.: 2003-10-3
Molecular Formula: C9H6BrF3O
Formula Weight: 267.04
Physical Form: Fused solid
Appearance: Pale yellow
Melting Point: 20-23°
Boiling Point: 69-72°
MDL No.: MFCD03094283
Formula:
C₉H₆BrF₃O Boiling Pt: 69…72 °C Melting Pt: 20…23 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094283 CAS Number: 2003-10-3 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...