(Trifluoromethyl)trimethylsilane 0.5 M in THF

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: trimethyl(trifluoromethyl)silane

H31520-AA H31520-AC H31520-AE
AAH31520-AAEA 30.5 USD
AAH31520-AA AAH31520-AC AAH31520-AE
(Trifluoromethyl)trimethylsilane 0.5 M in THF
(Trifluoromethyl)trimethylsilane

CAS No.: 81290-20-2 Molecular Formula: C4H9F3Si Formula Weight: 142.20 Flash Point: 17°(63°F) Boiling Point: 40° UN No.: 1993 MDL No.: MFCD00145454

Formula: C₄H₉F₃Si
MW: 142.2 g/mol
Boiling Pt: 40 °C
Density: 0.895
Flash Pt: 17 °C (63 °F)
Storage Temperature: Ambient
MDL Number: MFCD00145454
CAS Number: 81290-20-2
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR