4-Benzyloxy-2-fluorobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Benzyloxy)-2-fluorobenzoic acid

H31488-06 H31488-03
AAH31488-06EA 77.3 USD
AAH31488-06 AAH31488-03
4-Benzyloxy-2-fluorobenzoic acid 98%
4-Benzyloxy-2-fluorobenzoic acid

Purity: 98% CAS No.: 114045-96-4 Molecular Formula: C14H11FO3 Formula Weight: 246.24 Melting Point: 165-168° MDL No.: MFCD12407082

Formula: C₁₄H₁₁FO₃
MW: 246.24 g/mol
Melting Pt: 165…168 °C
Storage Temperature: Ambient
MDL Number: MFCD12407082
CAS Number: 114045-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR