4,4'-Dinonyl-2,2'-bipyridyl 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4,4′-Dinonyl-2,2′-dipyridyl, 4,4'-Dinonyl-2,2'-bipyridine, 4,4'-Di-n-nonyl-2,2'-bipyridine
H31476-03
H31476-06
AAH31476-03EA
61.4
USD
AAH31476-03
AAH31476-06
4,4'-Dinonyl-2,2'-bipyridyl 97%
4,4'-Dinonyl-2,2'-bipyridyl
Purity: 97%
CAS No.: 142646-58-0
Molecular Formula: C28H44N2
Formula Weight: 408.67
Melting Point: 61-63°
MDL No.: MFCD00800915
Formula:
C₂₈H₄₄N₂ MW: 408.67 g/mol Melting Pt: 61…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00800915 CAS Number: 142646-58-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...