3-(1,4-Cyclohexadien-1-yl)-1-propanol 97% stabilised
Supplier: Thermo Scientific Chemicals
Synonyms:
3-(Cyclohexa-1,4-dien-1-yl)propan-1-ol
H31465-03
H31465-06
AAH31465-03EA
83.02
USD
AAH31465-03
AAH31465-06
3-(1,4-Cyclohexadien-1-yl)-1-propanol 97% stabilised
3-(1,4-Cyclohexadien-1-yl)-1-propanol
Purity: 97%
CAS No.: 87151-66-4
Molecular Formula: C9H14O
Formula Weight: 138.21
Storage Temperature: -30°C to -10°C
Formula:
C₉H₁₄O MW: 138.21 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12407081 CAS Number: 87151-66-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...