Ethyl-2-cyano-2-methylpropionate 97%

Supplier: Thermo Scientific Chemicals
Danger

H31432-18 H31432-09
AAH31432-18EA 152 USD
AAH31432-18 AAH31432-09
Ethyl-2-cyano-2-methylpropionate 97%
Ethyl-2-cyano-2-methylpropionate

Purity: 97% CAS No.: 1572-98-1 Molecular Formula: C7H11NO2 Formula Weight: 141.17 Boiling Point: 185° MDL No.: MFCD00034667

Formula: C₇H₁₁NO₂
Boiling Pt: 185 °C
Density: 0.97
Storage Temperature: Ambient
MDL Number: MFCD00034667
CAS Number: 1572-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR