Acetoxy-2-propanone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H31346-AB
H31346-AD
AAH31346-ABEA
95.3
USD
AAH31346-AB
AAH31346-AD
Acetoxy-2-propanone 97%
Acetoxy-2-propanone
Purity: 97%
CAS No.: 592-20-1
Molecular Formula: C5H8O3
Formula Weight: 116.12
Flash Point: 71°(160°F)
Refractive Index: 1.4170
Boiling Point: 174-176°
MDL No.: MFCD00042848
Formula:
C₅H₈O₃ Boiling Pt: 174…176 °C Density: 1.075 Flash Pt: 71 °C(160 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042848 CAS Number: 592-20-1 EINECS: 209-746-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...