Boc-N-methyl-D-alanine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Boc-N-Me-Ala-OH, N-Boc-N-methyl-L-alanine

H31317-03 H31317-06
AAH31317-03EA 75.9 USD
AAH31317-03 AAH31317-06
Boc-N-methyl-D-alanine 98%
Boc-N-methyl-D-alanine

Purity: 98% CAS No.: 16948-16-6 Molecular Formula: C9H17NO4 Formula Weight: 203.24 Odor: Odorless Melting Point: 88-92° Optical Rotation: -31° (c=1 in ethanol) MDL No.: MFCD00037242

Formula: C₉H₁₇NO₄
MW: 203.2 g/mol
Melting Pt: 88…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00037242
CAS Number: 16948-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR