2,2'-Bis(diphenylphosphino)biphenyl 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
BIPHEP, 2,2'-Bis(diphenylphosphino)-1,1'-biphenyl
H31306-06
H31306-MD
H31306-03
AAH31306-06EA
622
USD
AAH31306-06
AAH31306-MD
AAH31306-03
2,2'-Bis(diphenylphosphino)biphenyl 98%
2,2'-Bis(diphenylphosphino)biphenyl
Purity: 98%
CAS No.: 84783-64-2
Molecular Formula: C36H28P2
Formula Weight: 522.57
Melting Point: 211-215°
MDL No.: MFCD03094574
Formula:
C₃₆H₂₈P₂ Melting Pt: 211…215 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094574 CAS Number: 84783-64-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...