5-Methylfuran-2-boronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 4,4,5,5-Tetramethyl-2-(5-methylfuran-2-yl)-1,3,2-dioxaborolane, 2-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)furan, (5-Methylfuran-2-yl)boronic acid Pinacol ester

H31282-03 H31282-06
AAH31282-03EA 24.6 USD
AAH31282-03 AAH31282-06
5-Methylfuran-2-boronic acid pinacol ester 97%
5-Methylfuran-2-boronic acid pinacol ester

Purity: 97% CAS No.: 338998-93-9 Molecular Formula: C11H17BO3 Formula Weight: 208.06 MDL No.: MFCD03094689

Formula: C₁₁H₁₇BO₃
MW: 208.06 g/mol
Density: 1,023
Storage Temperature: Ambient
MDL Number: MFCD03094689
CAS Number: 338998-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR