2-Cyclohexylethylboronic acid pinacol ester 96%
Supplier: Thermo Scientific Chemicals
H31156-03
H31156-06
AAH31156-03EA
67.6
USD
AAH31156-03
AAH31156-06
2-Cyclohexylethylboronic acid pinacol ester 96%
2-Cyclohexylethylboronic acid pinacol ester
Purity: 96%
CAS No.: 167692-95-7
Molecular Formula: C14H27BO2
Formula Weight: 238.18
MDL No.: MFCD12546193
Formula:
C₁₄H₂₇BO₂ Storage Temperature: Ambient |
MDL Number:
MFCD12546193 CAS Number: 167692-95-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...