(2-Hydroxylpropyl)-β-cyclodextrine, M.W. ∼1250-1480

Supplier: Thermo Scientific Chemicals

Synonyms: Hydroxypropyl Betadex, Hydroxypropyl-β-cyclodextrin, (2-Hydroxypropyl)-β-cyclodextrin, HP-β-CD

H31133-22 H31133-06 H31133-14
AAH31133-22EA 860.82 USD
AAH31133-22 AAH31133-06 AAH31133-14
(2-Hydroxylpropyl)-β-cyclodextrine, M.W. ∼1250-1480
(2-Hydroxylpropyl)-β-cyclodextrine

CAS No.: 128446-35-5 Melting Point: 305° MDL No.: MFCD00069372

Formula: C₆₃H₁₁₂O₄₂
Melting Pt: 305 °C
Storage Temperature: Ambient
MDL Number: MFCD16621721
CAS Number: 128446-35-5
EINECS: 420-920-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR