trans-4-Aminocyclohexanecarboxylic acid hydrochloride 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H31114-06
H31114-03
AAH31114-06EA
232
USD
AAH31114-06
AAH31114-03
trans-4-Aminocyclohexanecarboxylic acid hydrochloride 96%
trans-4-Aminocyclohexanecarboxylic acid hydrochloride
Purity: 96%
CAS No.: 27960-59-4
Molecular Formula: C7H13NO2.HCl
Formula Weight: 179.65
Melting Point:
Formula:
C₇H₁₄ClNO₂ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD03844587 CAS Number: 27960-59-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...