4-Amino-3-chloropyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

H31093-03 H31093-06
AAH31093-03EA 155 USD
AAH31093-03 AAH31093-06
4-Amino-3-chloropyridine 97%
4-Amino-3-chloropyridine

Purity: 97% CAS No.: 19798-77-7 Molecular Formula: C5H5ClN2 Formula Weight: 128.56 Odor: Unpleasant Melting Point: 49-53° MDL No.: MFCD03984473

Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Melting Pt: 49…53 °C
Storage Temperature: Ambient
MDL Number: MFCD03984473
CAS Number: 19798-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR