N-Formylglycine 98%

Supplier: Thermo Scientific Chemicals

H31053-06 H31053-03
AAH31053-06EA 104 USD
AAH31053-06 AAH31053-03
N-Formylglycine 98%
N-Formylglycine

Purity: 98% CAS No.: 2491-15-8 Molecular Formula: C3H5NO3 Formula Weight: 103.08 Storage Temperature: 2°C to 8°C Melting Point: 147-151° MDL No.: MFCD00037360

Formula: C₃H₅NO₃
Melting Pt: 147…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00037360
CAS Number: 2491-15-8
EINECS: 219-649-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR