3-Amino-1-methyl-1H-pyrazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Methyl-1H-pyrazol-3-amine, 3-Amino-1-methylpyrazole, 1-Methyl-1H-pyrazol-3-ylamine, 3-Amino-1-methyl-1H-pyrazole
H30935-03
H30935-06
AAH30935-03EA
20.5
USD
AAH30935-03
AAH30935-06
3-Amino-1-methyl-1H-pyrazole 97%
3-Amino-1-methyl-1H-pyrazole
Purity: 97%
CAS No.: 1904-31-0
Molecular Formula: C4H7N3
Formula Weight: 97.12
Boiling Point: 90-92°/1mm
UN No.: 2735
MDL No.: MFCD00466340
Formula:
C₄H₇N₃ MW: 97.12 g/mol Boiling Pt: 90…92 °C/1 mm Storage Temperature: Ambient |
MDL Number:
MFCD00466340 CAS Number: 1904-31-0 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...