2-Bromo-6-fluoropyridine 97%

Supplier: Thermo Scientific Chemicals
Danger

H30924-03 H30924-06
AAH30924-03EA 103 USD
AAH30924-03 AAH30924-06
2-Bromo-6-fluoropyridine 97%
2-Bromo-6-fluoropyridine

Purity: 97% CAS No.: 144100-07-2 Molecular Formula: C5H3BrFN Formula Weight: 175.99 Melting Point: 30-34° UN No.: 2811 MDL No.: MFCD03095258

Formula: C₅H₃BrFN
Melting Pt: 30…34 °C
Storage Temperature: Ambient
MDL Number: MFCD03095258
CAS Number: 144100-07-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR