D-Prolinamide 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-D-Pro-NH₂, (R)-2-Pyrrolidinecarboxamide, H-D-Pro-NH2
H30901-03
H30901-06
AAH30901-03EA
67.9
USD
AAH30901-03
AAH30901-06
D-Prolinamide 98%
D-Prolinamide
Purity: 98%
CAS No.: 62937-45-5
Molecular Formula: C5H10N2O
Formula Weight: 114.15
Melting Point: 100-103°
Optical Rotation: +107° (c=2 in ethanol)
MDL No.: MFCD00153457
Formula:
C₅H₁₀N₂O MW: 114.15 g/mol Melting Pt: 100…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153457 CAS Number: 62937-45-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...