3-Methyl-4-nitro-1H-pyrazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Methyl-4-nitropyrazole
H30860-14
H30860-06
AAH30860-14EA
245
USD
AAH30860-14
AAH30860-06
3-Methyl-4-nitro-1H-pyrazole 97%
3-Methyl-4-nitro-1H-pyrazole
Purity: 97%
CAS No.: 5334-39-4
Molecular Formula: C4H5N3O2
Formula Weight: 127.10
Melting Point: 131-134°
Boiling Point: 325°
MDL No.: MFCD00037864
Formula:
C4H5N3O2 MW: 127.10 g/mol Boiling Pt: 325 °C Melting Pt: 131…134 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037864 CAS Number: 5334-39-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...