4-(Boc-amino)pyridine 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Boc-Amino)pyridine

H30820-14 H30820-06
AAH30820-14EA 123 USD
AAH30820-14 AAH30820-06
4-(Boc-amino)pyridine 95%
4-(Boc-amino)pyridine

Purity: 95% CAS No.: 98400-69-2 Molecular Formula: C10H14N2O2 Formula Weight: 194.23 Melting Point: 147-150° MDL No.: MFCD02179232

Formula: C₁₀H₁₄N₂O₂
MW: 194.23 g/mol
Melting Pt: 147…150 °C
Storage Temperature: Ambient
MDL Number: MFCD02179232
CAS Number: 98400-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR