4-Amino-1-boc-piperidine 97%

Supplier: Thermo Scientific Chemicals
Danger

H30779-03 H30779-06
AAH30779-03EA 58.8 USD
AAH30779-03 AAH30779-06
4-Amino-1-boc-piperidine 97%
4-Amino-1-Boc-piperidine

Purity: 97% CAS No.: 87120-72-7 Molecular Formula: C10H20N2O2 Formula Weight: 200.28 Melting Point: 44-48° UN No.: 3259 MDL No.: MFCD01076201

Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Melting Pt: 44…48 °C
Storage Temperature: Ambient
MDL Number: MFCD01076201
CAS Number: 87120-72-7
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR